3-Chloro-4-(dimethylsulphamoyl)benzoic acid

Supplier: Apollo Scientific
Warning

OR52991-1G OR52991-250MG
APOSOR52991-1GEA 575 EUR
APOSOR52991-1G APOSOR52991-250MG
3-Chloro-4-(dimethylsulphamoyl)benzoic acid
3-Chloro-4-(dimethylsulphamoyl)benzoic acid
Formula: C₉H₁₀ClNO₄S
MW: 263,698 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18426353
CAS Number: 1263276-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR