2-Ethynyl-3-methylpyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-Methylpyridin-2-yl)acetylene, 2-Ethynyl-3-picoline
OR53007-1G
OR53007-250MG
APOSOR53007-1GEA
654
EUR
APOSOR53007-1G
APOSOR53007-250MG
2-Ethynyl-3-methylpyridine
2-Ethynyl-3-methylpyridine
Formula:
C₈H₇N MW: 117.148 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13175242 CAS Number: 30413-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...