2-Ethynylpyrazine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(Pyrazin-2-yl)acetylene, 2-Ethynyl-1,4-diazine
OR53022-1G
OR53022-250MG
OR53022-5G
APOSOR53022-1GEA
290
EUR
APOSOR53022-1G
APOSOR53022-250MG
APOSOR53022-5G
2-Ethynylpyrazine 98%
2-Ethynylpyrazine
Formula:
C₆H₄N₂ MW: 104,109 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13189804 CAS Number: 153800-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...