4-Bromo-3-hydroxybenzonitrile 98%

Supplier: Apollo Scientific

OR53056-100MG OR53056-250MG OR53056-1G
APOSOR53056-100MGEA 87 EUR
APOSOR53056-100MG APOSOR53056-250MG APOSOR53056-1G
4-Bromo-3-hydroxybenzonitrile 98%
4-Bromo-3-hydroxybenzonitrile
Formula: C₇H₄BrNO
MW: 198,017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999194
CAS Number: 916213-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR