6-Amino-2-ethyl-1,3-benzothiazole

Supplier: Apollo Scientific
Warning

OR53120-1G OR53120-250MG
APOSOR53120-1GEA 677 EUR
APOSOR53120-1G APOSOR53120-250MG
6-Amino-2-ethyl-1,3-benzothiazole
6-Amino-2-ethyl-1,3-benzothiazole
Formula: C₉H₁₀N₂S
MW: 178.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10686567
CAS Number: 17142-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR