2-Bromo-6-iodophenol 95%

Supplier: Apollo Scientific
Danger

OR54421-1G OR54421-250MG OR54421-5G
APOSOR54421-1GEA 253 EUR
APOSOR54421-1G APOSOR54421-250MG APOSOR54421-5G
2-Bromo-6-iodophenol 95%
2-Bromo-6-iodophenol
Formula: C₆H₄BrIO
MW: 298,904 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD16999870
CAS Number: 2040-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR