2-Bromo-3-nitrobenzonitrile

Supplier: Apollo Scientific
Warning

OR54427-1G OR54427-250MG
APOSOR54427-1GEA 609 EUR
APOSOR54427-1G APOSOR54427-250MG
2-Bromo-3-nitrobenzonitrile
2-Bromo-3-nitrobenzonitrile
Formula: C₇H₃BrN₂O₂
MW: 227,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392274
CAS Number: 90407-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR