7-(Aminomethyl)benzo[b]thiophene hydrochloride

Supplier: Apollo Scientific
Danger

Synonyms: (1-Benzothien-7-yl)methylamine hydrochloride

OR54443-1G OR54443-250MG
APOSOR54443-1GEA 811 EUR
APOSOR54443-1G APOSOR54443-250MG
7-(Aminomethyl)benzo[b]thiophene hydrochloride
7-(Aminomethyl)benzo[b]thiophene hydrochloride
Formula: C₉H₉NS·ClH
MW: 199,7 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27924152
CAS Number: 870562-96-2
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR