4-Bromo-5-chloro-2-methoxyaniline 95+%

Supplier: Apollo Scientific
Warning

OR54520-25G OR54520-5G
APOSOR54520-25GEA 134 EUR
APOSOR54520-25G APOSOR54520-5G
4-Bromo-5-chloro-2-methoxyaniline 95+%
4-Bromo-5-chloro-2-methoxyaniline
Formula: C₇H₇BrClNO
Storage Temperature: Ambient
MDL Number: MFCD09878150
CAS Number: 102170-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR