3,4-Dimethyloctane 95%

Supplier: Apollo Scientific
Warning

OR54624-1G OR54624-250MG
APOSOR54624-1GEA 186 EUR
APOSOR54624-1G APOSOR54624-250MG
3,4-Dimethyloctane 95%
3,4-Dimethyloctane
Formula: C₁₀H₂₂
MW: 142,282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30534471
CAS Number: 15869-92-8
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR