7-Bromo-2-iodo-1H-indole 95%
Supplier: Apollo Scientific
|
Warning
|
OR54695-1G
OR54695-250MG
APOSOR54695-1GEA
655
EUR
APOSOR54695-1G
APOSOR54695-250MG
7-Bromo-2-iodo-1H-indole 95%
7-Bromo-2-iodo-1H-indole
Formula:
C₈H₅BrIN MW: 321,94 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD32177106 CAS Number: 1904650-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...