(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid 95+%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-2,2'-((5-Amino-1-carboxypentyl)azanediyl)diacetic acid, Nα,Nα-Bis(carboxymethyl)-L-lysine

OR55124-5G
APOSOR55124-5GEA 97.3 EUR
APOSOR55124-5G
(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid 95+%
(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid
Formula: C₁₀H₁₈N₂O₆
MW: 262,26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237437
CAS Number: 113231-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR