Dibenzo[b,d]furan-4-ol

Supplier: Apollo Scientific
Warning

OR55145-1G OR55145-250MG
APOSOR55145-1GEA 78.3 EUR
APOSOR55145-1G APOSOR55145-250MG
Dibenzo[b,d]furan-4-ol
Dibenzo[b,d]furan-4-ol
Formula: C₁₂H₈O₂
MW: 184.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092287
CAS Number: 19261-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR