3-Bromo-4-nitrothiophenol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-4-nitrobenzenethiol
OR55174-1G
OR55174-250MG
APOSOR55174-1GEA
480
EUR
APOSOR55174-1G
APOSOR55174-250MG
3-Bromo-4-nitrothiophenol
3-Bromo-4-nitrothiophenol
Formula:
C₆H₄BrNO₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD22481215 CAS Number: 1823075-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...