1-Cyclopentyl-1,2,3-benzotriazole-5-carboxylic acid 98%

Supplier: Apollo Scientific
Warning

OR55243-1G OR55243-25G OR55243-5G
APOSOR55243-1GEA 161 EUR
APOSOR55243-1G APOSOR55243-25G APOSOR55243-5G
1-Cyclopentyl-1,2,3-benzotriazole-5-carboxylic acid 98%
1-Cyclopentyl-1,2,3-benzotriazole-5-carboxylic acid
Formula: C₁₂H₁₃N₃O₂
MW: 231,251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031748
CAS Number: 691363-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR