3-Methylbenzo[b]furan-2-carboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-methyl-1-benzofuran-2-carbaldehyde, 3-Methylbenzofuran-2-carbaldehyde
OR55307-250MG
OR55307-1G
APOSOR55307-250MGEA
74.2
EUR
APOSOR55307-250MG
APOSOR55307-1G
3-Methylbenzo[b]furan-2-carboxaldehyde
3-Methylbenzo[b]furan-2-carboxaldehyde
Formula:
C₁₀H₈O₂ MW: 160,169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01459584 CAS Number: 1199-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...