Methyl 3-(chloromethyl)benzoate 98%
Supplier: Apollo Scientific
OR55315-1G
OR55315-5G
APOSOR55315-1GEA
226
EUR
APOSOR55315-1G
APOSOR55315-5G
Methyl 3-(chloromethyl)benzoate 98%
Methyl 3-(chloromethyl)benzoate
Formula:
C₉H₉ClO₂ MW: 184,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00157184 CAS Number: 34040-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...