5-Butyl-1H-benzotriazole 98%
Supplier: Apollo Scientific
Synonyms:
5-Butyl-1H-benzo[d][1,2,3]triazole
OR55319-1G
OR55319-5G
APOSOR55319-1GEA
127
EUR
APOSOR55319-1G
APOSOR55319-5G
5-Butyl-1H-benzotriazole 98%
5-Butyl-1H-benzotriazole
Formula:
C₁₀H₁₃N₃ MW: 175,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22581692 CAS Number: 3663-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...