Methyl 2-amino-6-nitrobenzoate 97+%

Supplier: Apollo Scientific
Warning

OR55411-1G OR55411-5G
APOSOR55411-1GEA 139 EUR
APOSOR55411-1G APOSOR55411-5G
Methyl 2-amino-6-nitrobenzoate 97+%
Methyl 2-amino-6-nitrobenzoate
Formula: C₈H₈N₂O₄
MW: 196,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01851258
CAS Number: 57113-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR