2-Amino-6-nitrobenzaldehyde, Tech

Supplier: Apollo Scientific
Warning

OR55412-1G OR55412-250MG OR55412-5G
APOSOR55412-1GEA 394 EUR
APOSOR55412-1G APOSOR55412-250MG APOSOR55412-5G
2-Amino-6-nitrobenzaldehyde, Tech
2-Amino-6-nitrobenzaldehyde
Formula: C₇H₆N₂O₃
MW: 166,134 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18389895
CAS Number: 130133-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR