(2S)-2-(Methylamino)propan-1-ol 95%
Supplier: Apollo Scientific
|
Danger
|
OR55461-100MG
OR55461-1G
OR55461-250MG
APOSOR55461-100MGEA
139
EUR
APOSOR55461-100MG
APOSOR55461-1G
APOSOR55461-250MG
(2S)-2-(Methylamino)propan-1-ol 95%
(2S)-2-(Methylamino)propan-1-ol
Formula:
C₄H₁₁NO MW: 89,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07367992 CAS Number: 40916-73-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...