6-Bromoisoquinoline-1-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

OR55468-1G OR55468-5G
APOSOR55468-1GEA 619 EUR
APOSOR55468-1G APOSOR55468-5G
6-Bromoisoquinoline-1-carboxylic acid 97%
6-Bromoisoquinoline-1-carboxylic acid
Formula: C₁₀H₆BrNO₂
MW: 252,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257155
CAS Number: 1256806-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR