4-Isopropoxybenzonitrile 98%
Supplier: Apollo Scientific
OR55478-1G
OR55478-25G
OR55478-5G
APOSOR55478-1GEA
132
EUR
APOSOR55478-1G
APOSOR55478-25G
APOSOR55478-5G
4-Isopropoxybenzonitrile 98%
4-Isopropoxybenzonitrile
Formula:
C₁₀H₁₁NO MW: 161.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09929011 CAS Number: 91949-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...