4-Nitroindoline 95%
Supplier: Apollo Scientific
|
Warning
|
OR55492-1G
OR55492-250MG
APOSOR55492-1GEA
722
EUR
APOSOR55492-1G
APOSOR55492-250MG
4-Nitroindoline 95%
4-Nitroindoline
Formula:
C₈H₈N₂O₂ MW: 164.161 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07371624 CAS Number: 84807-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...