3-(Oxan-4-yl)-1,2,4-thiadiazol-5-amine 95%
Supplier: Apollo Scientific
|
Warning
|
OR55508-100MG
OR55508-1G
OR55508-250MG
APOSOR55508-100MGEA
510
EUR
APOSOR55508-100MG
APOSOR55508-1G
APOSOR55508-250MG
3-(Oxan-4-yl)-1,2,4-thiadiazol-5-amine 95%
3-(Oxan-4-yl)-1,2,4-thiadiazol-5-amine
Formula:
C₇H₁₁N₃OS MW: 185,247 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16745980 CAS Number: 1249828-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...