N-(4-Bromo-5-methyl-1,3-thiazol-2-yl)acetamide
Supplier: Apollo Scientific
|
Warning
|
OR55663-1G
OR55663-5G
APOSOR55663-1GEA
394
EUR
APOSOR55663-1G
APOSOR55663-5G
N-(4-Bromo-5-methyl-1,3-thiazol-2-yl)acetamide
N-(4-Bromo-5-methyl-1,3-thiazol-2-yl)acetamide
Formula:
C₆H₇BrN₂OS MW: 235,102 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29761446 CAS Number: 1934494-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...