1-(2-Cyclopropylphenyl)ethanone

Supplier: Apollo Scientific
Warning

OR55668-1G OR55668-250MG
APOSOR55668-1GEA 163 EUR
APOSOR55668-1G APOSOR55668-250MG
1-(2-Cyclopropylphenyl)ethanone
1-(2-Cyclopropylphenyl)ethanone
Formula: C₁₁H₁₂O
MW: 160.212 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06802722
CAS Number: 485402-63-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR