N-3-Thienylacetamide 99%

Supplier: Apollo Scientific
Warning

OR55684-10G OR55684-1G OR55684-5G
APOSOR55684-10GEA 328 EUR
APOSOR55684-10G APOSOR55684-1G APOSOR55684-5G
N-3-Thienylacetamide 99%
N-3-Thienylacetamide
Formula: C₆H₇NOS
MW: 141.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159570
CAS Number: 42602-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR