Ethyl 1-(4-bromophenyl)-1,2,4-triazole-3-carboxylate

Supplier: Apollo Scientific
Warning

OR55693-1G OR55693-250MG
APOSOR55693-1GEA 132 EUR
APOSOR55693-1G APOSOR55693-250MG
Ethyl 1-(4-bromophenyl)-1,2,4-triazole-3-carboxylate
Ethyl 1-(4-bromophenyl)-1,2,4-triazole-3-carboxylate
Formula: C₁₁H₁₀BrN₃O₂
MW: 296,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29905003
CAS Number: 1678521-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR