Ethyl 1-(ethoxymethyl)-1H-imidazole-4-carboxylate

Supplier: Apollo Scientific

OR59365-1G OR59365-250MG
APOSOR59365-1GEA 430 EUR
APOSOR59365-1G APOSOR59365-250MG
Ethyl 1-(ethoxymethyl)-1H-imidazole-4-carboxylate
Ethyl 1-(ethoxymethyl)-1H-imidazole-4-carboxylate
Formula: C₉H₁₄N₂O₃
MW: 198,219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878405
CAS Number: 957062-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR