N-[(5-Methyl-1,2-oxazol-3-yl)carbamothioyl]benzamide 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Benzoyl-3-(5-methylisoxazol-3-yl)thiourea
OR5954-1G
OR5954-5G
APOSOR5954-1GEA
51
EUR
APOSOR5954-1G
APOSOR5954-5G
N-[(5-Methyl-1,2-oxazol-3-yl)carbamothioyl]benzamide 97%
N-[(5-Methyl-1,2-oxazol-3-yl)carbamothioyl]benzamide
Formula:
C₁₂H₁₁N₃O₂S MW: 261,3 g/mol Melting Pt: 144…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126665 CAS Number: 85156-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...