Thiazol-2-ylmethanamine

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Aminomethyl)-1,3-thiazole, (1,3-Thiazol-2-yl)methylamine

OR59828-1G OR59828-250MG
APOSOR59828-1GEA 78.3 EUR
APOSOR59828-1G APOSOR59828-250MG
Thiazol-2-ylmethanamine
Thiazol-2-ylmethanamine
Formula: C₄H₆N₂S
MW: 114,17 g/mol
Density: 1,204 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD02854204
CAS Number: 55661-33-1
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR