2-Chloro-7,8-dihydropteridin-6(5H)-one 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-6-oxo-5,6,7,8-tetrahydropteridine
OR59870-1G
OR59870-250MG
APOSOR59870-1GEA
415
EUR
APOSOR59870-1G
APOSOR59870-250MG
2-Chloro-7,8-dihydropteridin-6(5H)-one 95+%
2-Chloro-7,8-dihydropteridin-6(5H)-one
Formula:
C₆H₅ClN₄O MW: 184,583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08062762 CAS Number: 944580-73-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...