5-(Fur-2-yl)pyridine-2-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Carboxy-5-(fur-2-yl)pyridine, 5-(Fur-2-yl)picolinic acid

OR59923-1G OR59923-250MG OR59923-50MG
APOSOR59923-1GEA 335 EUR
APOSOR59923-1G APOSOR59923-250MG APOSOR59923-50MG
5-(Fur-2-yl)pyridine-2-carboxylic acid 97%
5-(Fur-2-yl)pyridine-2-carboxylic acid
Formula: C₁₀H₇NO₃
MW: 189.167 g/mol
Melting Pt: 164…165 °C
Storage Temperature: Ambient
MDL Number: MFCD10700052
CAS Number: 930110-99-9
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR