3-Bromo-6-chloropyridine-2-carbonitrile 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Bromo-6-chloropicolinonitrile, 3-Bromo-6-chloro-2-cyanopyridine
OR60048-1G
OR60048-5G
APOSOR60048-1GEA
40.6
EUR
APOSOR60048-1G
APOSOR60048-5G
3-Bromo-6-chloropyridine-2-carbonitrile 97%
3-Bromo-6-chloropyridine-2-carbonitrile
Formula:
C₆H₂BrClN₂ MW: 217,45 g/mol Boiling Pt: 111…113 °C (0,15 mm Hg) Melting Pt: 87…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD10000884 CAS Number: 1053659-39-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...