5-Bromo-2-nitrotoluene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-methylnitrobenzene, 4-Bromo-2-methyl-1-nitrobenzene

OR60239-25G OR60239-5G OR60239-1G
APOSOR60239-25GEA 50.9 EUR
APOSOR60239-25G APOSOR60239-5G APOSOR60239-1G
5-Bromo-2-nitrotoluene 98%
5-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.032 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00137824
CAS Number: 52414-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR