3'-Iodo-2'-methylacetophenone 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(3-Iodo-2-methylphenyl)ethan-1-one), 1-(3-Iodo-2-methylphenyl)ethanone
OR60262-1G
OR60262-250MG
APOSOR60262-1GEA
299
EUR
APOSOR60262-1G
APOSOR60262-250MG
3'-Iodo-2'-methylacetophenone 95%
3'-Iodo-2'-methylacetophenone
Formula:
C₉H₉IO MW: 260,072 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22573017 CAS Number: 52164-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...