3-[(3-Methoxypropyl)carbamoyl]benzeneboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: N-(3-Methoxypropyl) 3-boronobenzamide

OR6101-1G OR6101-5G
APOSOR6101-1GEA 216 EUR
APOSOR6101-1G APOSOR6101-5G
3-[(3-Methoxypropyl)carbamoyl]benzeneboronic acid 98%
3-[(3-Methoxypropyl)carbamoyl]benzeneboronic acid
Formula: C₁₁H₁₆BNO₄
MW: 237,06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09258755
CAS Number: 957061-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR