4-[2-(1H-Pyrazol-1-yl)ethoxy]benzeneboronic acid 96%
Supplier: Apollo Scientific
OR6109-1G
OR6109-250MG
APOSOR6109-1GEA
786
EUR
APOSOR6109-1G
APOSOR6109-250MG
4-[2-(1H-Pyrazol-1-yl)ethoxy]benzeneboronic acid 96%
4-[2-(1H-Pyrazol-1-yl)ethoxy]benzeneboronic acid
Formula:
C₁₁H₁₃BN₂O₃ MW: 232,044 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258754 CAS Number: 957061-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...