2-(2-Ethoxyphenoxy)ethylamine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(2-Aminoethoxy)-2-ethoxybenzene, 2-(2-Ethoxyphenoxy)ethanamine
OR61209-1G
OR61209-250MG
APOSOR61209-1GEA
299
EUR
APOSOR61209-1G
APOSOR61209-250MG
2-(2-Ethoxyphenoxy)ethylamine
2-(2-Ethoxyphenoxy)ethylamine
Formula:
C₁₀H₁₅NO₂ MW: 181.232 g/mol Boiling Pt: 94…96 °C (1 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06657756 CAS Number: 6781-17-5 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...