2-Bromo-N4-methylpyridine-3,4-diamine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-2-bromo-4-(methylamino)pyridine

OR61613-1G OR61613-250MG
APOSOR61613-1GEA 0 EUR
APOSOR61613-1G APOSOR61613-250MG
2-Bromo-N4-methylpyridine-3,4-diamine
2-Bromo-N4-methylpyridine-3,4-diamine
Formula: C₆H₈BrN₃
MW: 202,052 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415287
CAS Number: 1396554-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR