6-Bromo-3-methyl-1H-indole

Supplier: Apollo Scientific
Warning

OR62080-1G OR62080-5G
APOSOR62080-1GEA 89.9 EUR
APOSOR62080-1G APOSOR62080-5G
6-Bromo-3-methyl-1H-indole
6-Bromo-3-methyl-1H-indole
Formula: C₉H₈BrN
MW: 210.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834853
CAS Number: 1219741-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR