4-Aminophenyl-β-D-mannopyranoside hydrochloride
Supplier: Apollo Scientific
OR6250T-100MG
OR6250T-50MG
APOSOR6250T-100MGEA
662
EUR
APOSOR6250T-100MG
APOSOR6250T-50MG
4-Aminophenyl-β-D-mannopyranoside hydrochloride
4-Aminophenyl-β-D-mannopyranoside hydrochloride
Formula:
C₁₂H₁₇NO₆·ClH MW: 307,727 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00190750 CAS Number: 210049-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...