Ethyl 2-cyano-3-[4-(dimethylamino)phenyl]acrylate
Supplier: Apollo Scientific
|
Warning
|
OR6291-1G
OR6291-2.5G
APOSOR6291-1GEA
232
EUR
APOSOR6291-1G
APOSOR6291-2.5G
Ethyl 2-cyano-3-[4-(dimethylamino)phenyl]acrylate
Ethyl 2-cyano-3-[4-(dimethylamino)phenyl]acrylate
Formula:
C₁₄H₁₆N₂O₂ Melting Pt: 123…125 °C Density: 1,23 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00026847 CAS Number: 1886-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...