Thieno[2,3-b]pyridin-2-ylmethanol 97%
Supplier: Apollo Scientific
|
Warning
|
OR6750-1G
OR6750-250MG
APOSOR6750-1GEA
190
EUR
APOSOR6750-1G
APOSOR6750-250MG
Thieno[2,3-b]pyridin-2-ylmethanol 97%
Thieno[2,3-b]pyridin-2-ylmethanol
Formula:
C₈H₇NOS MW: 165,212 g/mol Melting Pt: 46,5…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD07772792 CAS Number: 131337-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...