1-[3-(Pyrrolidin-1-yl)prop-1-yl]piperazine 97%

Supplier: Apollo Scientific
Danger

OR6891-1G OR6891-500MG
APOSOR6891-1GEA 132 EUR
APOSOR6891-1G APOSOR6891-500MG
1-[3-(Pyrrolidin-1-yl)prop-1-yl]piperazine 97%
1-[3-(Pyrrolidin-1-yl)prop-1-yl]piperazine
Formula: C₁₁H₂₃N₃
Boiling Pt: 106…108 °C (0,2 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01320895
CAS Number: 224309-80-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR