1-[3-(Dipropylamino)prop-1-yl]piperazine 97%
Supplier: Apollo Scientific
|
Warning
|
OR6917-1G
OR6917-250MG
APOSOR6917-1GEA
17.6
EUR
APOSOR6917-1G
APOSOR6917-250MG
1-[3-(Dipropylamino)prop-1-yl]piperazine 97%
1-[3-(Dipropylamino)prop-1-yl]piperazine
Formula:
C₁₃H₂₉N₃ Boiling Pt: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075183 CAS Number: 827614-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...