7-Bromo-1,3-benzoxazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Bromobenzo[d]oxazole
OR70006-250MG
OR70006-1G
OR70006-5G
APOSOR70006-250MGEA
29.2
EUR
APOSOR70006-250MG
APOSOR70006-1G
APOSOR70006-5G
7-Bromo-1,3-benzoxazole
7-Bromo-1,3-benzoxazole
Formula:
C₇H₄BrNO MW: 198,017 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234619 CAS Number: 885270-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...