8-Azabicyclo[3.2.1]octane hydrochloride 95+%

Supplier: Apollo Scientific

OR70094-100MG OR70094-250MG OR70094-1G
APOSOR70094-100MGEA 181 EUR
APOSOR70094-100MG APOSOR70094-250MG APOSOR70094-1G
8-Azabicyclo[3.2.1]octane hydrochloride 95+%
8-Azabicyclo[3.2.1]octane hydrochloride
Formula: C₇H₁₃N·ClH
MW: 147,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19982561
CAS Number: 6760-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR