4-Phenoxyphenethyl alcohol

Supplier: Apollo Scientific
Warning

OR70177-1G OR70177-5G
APOSOR70177-1GEA 812 EUR
APOSOR70177-1G APOSOR70177-5G
4-Phenoxyphenethyl alcohol
4-Phenoxyphenethyl alcohol
Formula: C₁₄H₁₄O₂
Boiling Pt: 142 °C (0,1 mm Hg)
Melting Pt: 14…15 °C
Density: 1,1378 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06201083
CAS Number: 52446-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR